Fmoc-DL-(2-bromophenyl)glycine

Supplier: Matrix Scientific

041569-1G 041569-5G
101852-306EA 547.69 USD
101852-306 101852-308
Fmoc-DL-(2-bromophenyl)glycine
Fmoc-DL-(2-bromophenyl)glycine

Hazard: Irritant

Formula: C₂₃H₁₈BrNO₄
MDL Number: MFCD09750503
CAS Number: 1219346-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR