Fmoc-D-Chg-OH

Supplier: Matrix Scientific

041567-1G 041567-5G
101852-298EA 187.69 USD
101852-298 101852-300
Fmoc-D-Chg-OH
Fmoc-D-Chg-OH

Hazard: Irritant

Formula: C₂₃H₂₅NO₄
MDL Number: MFCD00190877
CAS Number: 198543-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR