Fmoc-D-Tle-OH
Supplier: Matrix Scientific
Synonyms:
Fmoc-tBu-D-Gly-OH
041566-1G
041566-5G
101852-294EA
598.46
USD
101852-294
101852-296
Fmoc-D-Tle-OH
Fmoc-D-Tle-OH
Hazard: Irritant
Formula:
C₂₁H₂₃NO₄ MW: 353.418 g/mol |
MDL Number:
MFCD00065650 CAS Number: 198543-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...