Fmoc-D-Abu-OH

Supplier: Matrix Scientific

Synonyms: (R)-2-(Fmoc-amino)butyric acid

041565-1G 041565-5G
101852-290EA 190.77 USD
101852-290 101852-292
Fmoc-D-Abu-OH
Fmoc-D-Abu-OH

Hazard: Irritant

Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
MDL Number: MFCD00270336
CAS Number: 170642-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR