Fmoc-D-Dab-OH
Supplier: Matrix Scientific
Synonyms:
(R)-2-(Fmoc-amino)-4-aminobutyric acid
041564-1G
041564-5G
101852-286EA
586.15
USD
101852-286
101852-288
Fmoc-D-Dab-OH
Fmoc-D-Dab-OH
Hazard: Irritant
Formula:
C₁₉H₂₀N₂O₄ MW: 340.379 g/mol |
MDL Number:
MFCD00237018 CAS Number: 201484-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...