Fmoc-D-allylglycine

Supplier: Matrix Scientific

Synonyms: D-2-Allylglycine, N-FMOC protected, Fmoc-D-Gly(allyl)-OH

041562-1G 041562-5G
101852-278EA 56.92 USD
101852-278 101852-280
Fmoc-D-allylglycine
Fmoc-D-allylglycine

Hazard: Irritant

Formula: C₂₀H₁₉NO₄
MW: 337.38 g/mol
MDL Number: MFCD01311776
CAS Number: 170642-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR