N-Fmoc-D-norleucine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Nle-OH

041560-5G 041560-25G
101852-272EA 143.08 USD
101852-272 102833-114
N-Fmoc-D-norleucine
N-Fmoc-D-norleucine

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
MDL Number: MFCD00155639
CAS Number: 112883-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR