Fmoc-D-cyclopentylglycine
Supplier: Matrix Scientific
Synonyms:
Fmoc-D-Cpg-OH
041556-250MG
041556-1G
101852-254EA
436.92
USD
101852-254
101852-256
Fmoc-D-cyclopentylglycine
Fmoc-D-cyclopentylglycine
Hazard: Irritant
Formula:
C₂₂H₂₃NO₄ MW: 365.429 g/mol |
MDL Number:
MFCD01311778 CAS Number: 136555-16-3 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...