Fmoc-β-t-butyl-D-alanine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-tBu-Ala-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4,4-dimethylpentanoic acid

041554-250MG 041554-1G
101852-246EA 592.31 USD
101852-246 101852-248
Fmoc-β-t-butyl-D-alanine
Fmoc-beta-t-butyl-D-alanine

Hazard: Irritant

Formula: C₂₂H₂₅NO₄
MDL Number: MFCD01861369
CAS Number: 359766-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR