Fmoc-β-cyclopentyl-DL-alanine

Supplier: Matrix Scientific

041552-100MG 041552-500MG
101852-238EA 498.46 USD
101852-238 101852-240
Fmoc-β-cyclopentyl-DL-alanine
Fmoc-beta-cyclopentyl-DL-alanine

Hazard: Irritant

Formula: C₂₃H₂₅NO₄
MW: 379.46 g/mol
MDL Number: MFCD02682459
CAS Number: 1219422-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR