Fmoc-β-cyclopenten-1-yl-DL-alanine

Supplier: Matrix Scientific

041551-100MG 041551-500MG
101852-234EA 400 USD
101852-234 101852-236
Fmoc-β-cyclopenten-1-yl-DL-alanine
Fmoc-beta-cyclopenten-1-yl-DL-alanine

Hazard: Irritant

Formula: C₂₃H₂₃NO₄
MW: 377.44 g/mol
MDL Number: MFCD02682461
CAS Number: 1219148-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR