Fmoc-β-cyano-L-alanine

Supplier: Matrix Scientific

041548-1G 041548-5G
101852-220EA 547.69 USD
101852-220 101852-222
Fmoc-β-cyano-L-alanine
Fmoc-beta-cyano-L-alanine

Hazard: Irritant

Formula: C₁₉H₁₆N₂O₄
MDL Number: MFCD01861370
CAS Number: 127273-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR