Fmoc-D-4-pyridylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-4-Pal-OH

041545-1G 041545-5G
101852-208EA 349.23 USD
101852-208 101852-210
Fmoc-D-4-pyridylalanine
Fmoc-D-4-pyridylalanine

Hazard: Irritant

Formula: C₂₃H₂₀N₂O₄
MW: 388.43 g/mol
MDL Number: MFCD00672567
CAS Number: 205528-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR