Fmoc-3-(3-pyridyl)-L-alanine

Supplier: Matrix Scientific

Synonyms: Fmoc-3-Pal-OH, Fmoc-β-(3-pyridyl)-Ala-OH

041544-1G 041544-5G
101852-204EA 192.31 USD
101852-204 101852-206
Fmoc-3-(3-pyridyl)-L-alanine
Fmoc-3-(3-pyridyl)-L-alanine

Hazard: Irritant

Formula: C₂₃H₂₀N₂O₄
MW: 388.423 g/mol
MDL Number: MFCD00144887
CAS Number: 175453-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR