Fmoc-3-(3-pyridyl)-D-alanine
Supplier: Matrix Scientific
Synonyms:
Fmoc-β-(3-pyridyl)-D-Ala-OH, Fmoc-D-3-Pal-OH
041543-1G
041543-5G
101852-200EA
238.46
USD
101852-200
101852-202
Fmoc-3-(3-pyridyl)-D-alanine
Fmoc-3-(3-pyridyl)-D-alanine
Hazard: Irritant
Formula:
C₂₃H₂₀N₂O₄ MW: 388.423 g/mol |
MDL Number:
MFCD00080267 CAS Number: 142994-45-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...