Fmoc-β-(3-benzothienyl)-Ala-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-3-Ala(3-benzothienyl)-OH, Fmoc-β-(3-benzothienyl)-Ala-OH

041542-1G 041542-5G
101852-196EA 326.15 USD
101852-196 101852-198
Fmoc-β-(3-benzothienyl)-Ala-OH
Fmoc-beta-(3-benzothienyl)-Ala-OH

Hazard: Irritant

Formula: C₂₆H₂₁NO₄S
MDL Number: MFCD00672562
CAS Number: 177966-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR