Fmoc-β-(2-thienyl)-D-alanine
Supplier: Matrix Scientific
Synonyms:
(R)-N-Fmoc-2-thienylalanine, Fmoc-D-Thi-OH, Fmoc-D-3-(2-Thienyl)-Ala-OH, Fmoc-3-D-Ala(2-thienyl)-OH, Fmoc-β-(2-thienyl)-D-Ala-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(thiophen-2-yl)propanoic acid, Fmoc-D-2-thienylalanine
041539-1G
041539-5G
101852-184EA
224.62
USD
101852-184
101852-186
Fmoc-β-(2-thienyl)-D-alanine
Fmoc-β-(2-thienyl)-D-alanine
Hazard: Irritant
Formula:
C₂₂H₁₉NO₄S MW: 393.457 g/mol |
MDL Number:
MFCD00065677 CAS Number: 201532-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...