Fmoc-3-(1-naphthyl)-L-alanine

Supplier: Matrix Scientific

Synonyms: Fmoc-1-Nal-OH

041534-1G 041534-5G
101852-164EA 126.15 USD
101852-164 101852-166
Fmoc-3-(1-naphthyl)-L-alanine
Fmoc-3-(1-naphthyl)-L-alanine

Hazard: Irritant

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
MDL Number: MFCD00151914
CAS Number: 96402-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR