Fmoc-3-(1-naphthyl)-D-alanine
Supplier: Matrix Scientific
041533-1G
041533-5G
101852-160EA
224.62
USD
101852-160
101852-162
Fmoc-3-(1-naphthyl)-D-alanine
Fmoc-3-(1-naphthyl)-D-alanine
Hazard: Irritant
Formula:
C₂₈H₂₃NO₄ |
MDL Number:
MFCD00191200 CAS Number: 138774-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...