Fmoc-6-Ahx-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-ε-aminocaproic acid, Fmoc-ε-Acp-OH, 6-(Fmoc-amino)hexanoic acid

041529-1G 041529-5G
101852-144EA 18.46 USD
101852-144 101852-146
Fmoc-6-Ahx-OH
Fmoc-6-Ahx-OH

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
MW: 353.418 g/mol
MDL Number: MFCD00077045
CAS Number: 88574-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR