Fmoc-3-amino-3-(2-thienyl)propionic acid

Supplier: Matrix Scientific

041528-1G 041528-5G
101852-140EA 823.08 USD
101852-140 101852-142
Fmoc-3-amino-3-(2-thienyl)propionic acid
Fmoc-3-amino-3-(2-thienyl)propionic acid

Hazard: Irritant

Formula: C₂₂H₁₉NO₄S
MW: 393.47 g/mol
MDL Number: MFCD01863207
CAS Number: 282525-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR