Fmoc-(S)-3-amino-6-phenyl-5-hexenoic acid
Supplier: Matrix Scientific
041524-250MG
041524-1G
101852-124EA
412.31
USD
101852-124
101852-126
Fmoc-(S)-3-amino-6-phenyl-5-hexenoic acid
Fmoc-(S)-3-amino-6-phenyl-5-hexenoic acid
Hazard: Irritant
Formula:
C₂₇H₂₅NO₄ |
MDL Number:
MFCD01861005 CAS Number: 270596-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...