Fmoc-(S)-3-amino-5-phenylpentanoic acid

Supplier: Matrix Scientific

Synonyms: Fmoc-β-Nva(5-phenyl)-OH, (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-phenylpentanoic acid

041523-250MG 041523-1G
101852-120EA 275.38 USD
101852-120 101852-122
Fmoc-(S)-3-amino-5-phenylpentanoic acid
Fmoc-(S)-3-amino-5-phenylpentanoic acid

Hazard: Irritant

Formula: C₂₆H₂₅NO₄
MDL Number: MFCD01861029
CAS Number: 219967-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR