Fmoc-(S)-3-amino-5-hexynoic acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)hex-5-ynoic acid

041522-250MG 041522-1G
101852-116EA 461.54 USD
101852-116 101852-118
Fmoc-(S)-3-amino-5-hexynoic acid
Fmoc-(S)-3-amino-5-hexynoic acid

Hazard: Irritant

Formula: C₂₁H₁₉NO₄
MDL Number: MFCD01861008
CAS Number: 1217669-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR