Fmoc-(S)-3-amino-4-pentafluorophenylbutyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(perfluorophenyl)butanoic acid

041520-250MG 041520-1G
101852-108EA 412.31 USD
101852-108 101852-110
Fmoc-(S)-3-amino-4-pentafluorophenylbutyric acid
Fmoc-(S)-3-amino-4-pentafluorophenylbutyric acid

Hazard: Irritant

Formula: C₂₅H₁₈F₅NO₄
MW: 491.42 g/mol
MDL Number: MFCD01861039
CAS Number: 270063-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR