Fmoc-(S)-3-amino-4-(4-pyridyl)butyric acid

Supplier: Matrix Scientific

041518-250MG 041518-1G
101852-100EA 510.77 USD
101852-100 101852-102
Fmoc-(S)-3-amino-4-(4-pyridyl)butyric acid
Fmoc-(S)-3-amino-4-(4-pyridyl)butyric acid

Hazard: Irritant

Formula: C₂₄H₂₂N₂O₄
MDL Number: MFCD01861059
CAS Number: 270065-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR