Fmoc-(S)-3-amino-4-(4-nitrophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(4-nitrophenyl)butanoic acid

041517-250MG 041517-1G
101852-096EA 275.38 USD
101852-096 101852-098
Fmoc-(S)-3-amino-4-(4-nitrophenyl)butyric acid
Fmoc-(S)-3-amino-4-(4-nitrophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂N₂O₆
MW: 446.46 g/mol
MDL Number: MFCD01861018
CAS Number: 270062-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR