Fmoc-(S)-3-amino-4-(4-iodophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(4-iodophenyl)butanoic acid

041515-250MG 041515-1G
101852-088EA 275.38 USD
101852-088 101852-090
Fmoc-(S)-3-amino-4-(4-iodophenyl)butyric acid
Fmoc-(S)-3-amino-4-(4-iodophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂INO₄
MW: 527.36 g/mol
MDL Number: MFCD01861062
CAS Number: 270065-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR