Fmoc-(S)-3-amino-4-(4-fluorophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(4-fluorophenyl)butanoic acid

041514-250MG 041514-1G
101852-084EA 275.38 USD
101852-084 101852-086
Fmoc-(S)-3-amino-4-(4-fluorophenyl)butyric acid
Fmoc-(S)-3-amino-4-(4-fluorophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂FNO₄
MW: 419.46 g/mol
MDL Number: MFCD01861013
CAS Number: 270062-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR