Fmoc-(S)-3-amino-4-(4-cyanophenyl)butyric acid

Supplier: Matrix Scientific

041513-250MG 041513-1G
101852-080EA 412.31 USD
101852-080 101852-082
Fmoc-(S)-3-amino-4-(4-cyanophenyl)butyric acid
Fmoc-(S)-3-amino-4-(4-cyanophenyl)butyric acid

Hazard: Irritant

Formula: C₂₆H₂₂N₂O₄
MDL Number: MFCD01861080
CAS Number: 270065-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR