Fmoc-4-fluoro-L-phenylglycine
Supplier: Matrix Scientific
041510-1G
041510-5G
101852-068EA
1046.15
USD
101852-068
101852-070
Fmoc-4-fluoro-L-phenylglycine
Fmoc-4-fluoro-L-phenylglycine
Hazard: Irritant
Formula:
C₂₃H₁₈FNO₄ |
MDL Number:
MFCD01320845 CAS Number: 678988-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...