Fmoc-4-fluoro-D-phenylglycine
Supplier: Matrix Scientific
041509-1G
041509-5G
101852-064EA
1046.15
USD
101852-064
101852-066
Fmoc-4-fluoro-D-phenylglycine
Fmoc-4-fluoro-D-phenylglycine
Hazard: Irritant
Formula:
C₂₃H₁₈FNO₄ |
MDL Number:
MFCD01320876 CAS Number: 374791-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...