Fmoc-4,5-dehydro-l-leucine

Supplier: Matrix Scientific

Synonyms: Fmoc-4,5-dehydro-Leu-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methylpent-4-enoic acid

041508-250MG 041508-1G
101852-060EA 233.85 USD
101852-060 101852-062
Fmoc-4,5-dehydro-l-leucine
Fmoc-4,5-dehydro-l-leucine

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
MDL Number: MFCD00237651
CAS Number: 87720-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR