Fmoc-4,4,4-trifluoro-DL-valine

Supplier: Matrix Scientific

041507-100MG 041507-1G
101852-056EA 424.62 USD
101852-056 101852-058
Fmoc-4,4,4-trifluoro-DL-valine
Fmoc-4,4,4-trifluoro-DL-valine

Hazard: Irritant

Formula: C₂₀H₁₈F₃NO₄
MDL Number: MFCD02682486
CAS Number: 1219402-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR