Fmoc-3-fluoro-DL-valine

Supplier: Matrix Scientific

041506-100MG 041506-250MG
101852-052EA 1395.38 USD
101852-052 101852-054
Fmoc-3-fluoro-DL-valine
Fmoc-3-fluoro-DL-valine

Hazard: Irritant

Formula: C₂₀H₂₀FNO₄
MDL Number: MFCD02682474
CAS Number: 1219144-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR