Fmoc-3-amino-4,4,4-trifluorobutyric acid
Supplier: Matrix Scientific
Synonyms:
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4,4,4-trifluorobutanoic acid
041505-100MG
041505-250MG
041505-500MG
101852-046EA
350.81
USD
101852-046
101852-048
101852-050
Fmoc-3-amino-4,4,4-trifluorobutyric acid
Fmoc-3-amino-4,4,4-trifluorobutyric acid
Hazard: Irritant
Formula:
C₁₉H₁₆F₃NO₄ MW: 379.34 g/mol |
MDL Number:
MFCD02682449 CAS Number: 194471-86-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...