Fmoc-2-fluoro-DL-phenylglycine
Supplier: Matrix Scientific
Synonyms:
2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-2-(2-fluorophenyl)acetic acid
041504-1G
041504-5G
101852-042EA
412.31
USD
101852-042
101852-044
Fmoc-2-fluoro-DL-phenylglycine
Fmoc-2-fluoro-DL-phenylglycine
Hazard: Irritant
Formula:
C₂₃H₁₈FNO₄ MW: 391.4 g/mol |
MDL Number:
MFCD02682470 CAS Number: 433291-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...