Fmoc-2-amino-4,4,4-trifluorobutyric acid

Supplier: Matrix Scientific

041503-100MG 041503-500MG
101852-038EA 498.46 USD
101852-038 101852-040
Fmoc-2-amino-4,4,4-trifluorobutyric acid
Fmoc-2-amino-4,4,4-trifluorobutyric acid

Hazard: Irritant

Formula: C₁₉H₁₆F₃NO₄
MDL Number: MFCD02682447
CAS Number: 1219145-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR