Fmoc-2,4-dinitro-DL-phenylglycine

Supplier: Matrix Scientific

041501-100MG 041501-500MG
101852-030EA 275.38 USD
101852-030 101852-032
Fmoc-2,4-dinitro-DL-phenylglycine
Fmoc-2,4-dinitro-DL-phenylglycine

Hazard: Irritant

Formula: C₂₃H₁₇N₃O₈
MW: 463.41 g/mol
MDL Number: MFCD02682467
CAS Number: 1219389-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR