Fmoc-Dab(Boc)-OH

Supplier: Matrix Scientific

Synonyms: (2S)-2,4-Diaminobutanoic acid, 4-BOC, 2-FMOC protected

041500-1G 041500-5G
101852-026EA 43.08 USD
101852-026 101852-028
Fmoc-Dab(Boc)-OH
Fmoc-Dab(Boc)-OH

Hazard: Irritant

Formula: C₂₄H₂₈N₂O₆
MW: 440.496 g/mol
Melting Pt: 111…113 °C
MDL Number: MFCD00151941
CAS Number: 125238-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR