Fmoc-Dab(Alloc)-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-Dab(Aloc)-OH

041498-1G 041498-5G
101852-018EA 326.15 USD
101852-018 101852-020
Fmoc-Dab(Alloc)-OH
Fmoc-Dab(Alloc)-OH

Hazard: Irritant

Formula: C₂₃H₂₄N₂O₆
MW: 424.453 g/mol
MDL Number: MFCD00798634
CAS Number: 204316-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR