Fmoc-(N-γ-4-methyltrityl)-L-α,γ-diaminobutyric acid

Supplier: Matrix Scientific

Synonyms: Fmoc-Dab(Mtt)-OH

041497-1G 041497-5G
101852-014EA 686.15 USD
101852-014 101852-016
Fmoc-(N-γ-4-methyltrityl)-L-α,γ-diaminobutyric acid
Fmoc-(N-gamma-4-methyltrityl)-L-alpha,gamma-diaminobutyric acid

Hazard: Irritant

Formula: C₃₉H₃₆N₂O₄
MDL Number: MFCD02094096
CAS Number: 851392-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR