Fmoc-(N-γ-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-α,γ-diaminobutyric acid

Supplier: Matrix Scientific

Synonyms: Fmoc-Dab(Dde)-OH

041495-1G 041495-5G
101852-006EA 661.54 USD
101852-006 101852-008
Fmoc-(N-γ-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-α,γ-diaminobutyric acid
Fmoc-(N-gamma-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-alpha,gamma-diaminobutyric acid

Hazard: Irritant

Formula: C₂₉H₃₂N₂O₆
MDL Number: MFCD11111552
CAS Number: 235788-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR