Fmoc-(N-γ-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-α,γ-diaminobutyric acid

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Dab(Dde)-OH

041494-1G 041494-5G
101852-002EA 686.15 USD
101852-002 101852-004
Fmoc-(N-γ-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-α,γ-diaminobutyric acid
Fmoc-(N-gamma-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-alpha,gamma-diaminobutyric acid

Hazard: Irritant

Formula: C₂₉H₃₂N₂O₆
MW: 504.59 g/mol
MDL Number: MFCD09265180
CAS Number: 596797-14-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR