Fmoc-Dap(Boc)-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-(N-beta-boc)-L-alpha,beta-diaminopropionic acid, 3-(tert-Butoxycarbonyl)amino-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-alanine, (S)-3-(tert-Butoxycarbonylamino)-2-[(9H-fluoren-9-ylmethoxy)carbonylamino]propionic acid, Nα-Fmoc-Nβ-Boc-L-2,3-diaminopropionic acid, (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 3-(Boc-amino)-N-Fmoc-L-alanine

041493-1G 041493-5G
101851-998EA 210.77 USD
101851-998 101852-000
Fmoc-Dap(Boc)-OH
Fmoc-Dap(Boc)-OH

Hazard: Irritant

Formula: C₂₃H₂₆N₂O₆
MW: 426.47 g/mol
MDL Number: MFCD00235896
CAS Number: 162558-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR