Fmoc-D-Dap(Boc)-OH

Supplier: Matrix Scientific

Synonyms: (R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid

041492-1G 041492-5G
101851-994EA 469.23 USD
101851-994 101851-996
Fmoc-D-Dap(Boc)-OH
Fmoc-D-Dap(Boc)-OH

Hazard: Irritant

Formula: C₂₃H₂₆N₂O₆
MDL Number: MFCD00798648
CAS Number: 198544-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR