Fmoc-(N-β-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-α,β-diaminopropionic acid

Supplier: Matrix Scientific

Synonyms: Fmoc-Dap(Dde)-OH

041489-1G 041489-5G
101851-982EA 366.15 USD
101851-982 101851-984
Fmoc-(N-β-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-α,β-diaminopropionic acid
Fmoc-(N-beta-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-L-alpha,beta-diaminopropionic acid

Hazard: Irritant

Formula: C₂₈H₃₀N₂O₆
MW: 490.56 g/mol
MDL Number: MFCD01631974
CAS Number: 247127-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR