(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((2,4-dinitrophenyl)amino)propanoic acid
Supplier: Matrix Scientific
Synonyms:
Fmoc-Dap(Dnp)-OH
041487-1G
041487-5G
101851-974EA
443.08
USD
101851-974
101851-976
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((2,4-dinitrophenyl)amino)propanoic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((2,4-dinitrophenyl)amino)propanoic acid
Hazard: Irritant
Formula:
C₂₄H₂₀N₄O₈ MW: 492.444 g/mol |
MDL Number:
MFCD00798650 CAS Number: 140430-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...