(S)-2,4-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)butanoic acid

Supplier: Matrix Scientific

Synonyms: Fmoc-Dab(Fmoc)-OH, (S)-2,4-Bis(Fmoc-amino)butyric acid

041485-1G 041485-5G
101851-966EA 275.38 USD
101851-966 101851-968
(S)-2,4-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)butanoic acid
(S)-2,4-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)butanoic acid

Hazard: Irritant

Formula: C₃₄H₃₀N₂O₆
MDL Number: MFCD00237004
CAS Number: 201473-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR