Fmoc-Dap(Fmoc)-OH

Supplier: Matrix Scientific

Synonyms: Nα-Fmoc-3-(Fmoc-amino)-L-alanine, (S)-2,3-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propanoic acid

041484-1G 041484-5G
101851-962EA 275.38 USD
101851-962 101851-964
Fmoc-Dap(Fmoc)-OH
Fmoc-Dap(Fmoc)-OH

Hazard: Irritant

Formula: C₃₃H₂₈N₂O₆
MW: 548.595 g/mol
MDL Number: MFCD00237005
CAS Number: 201473-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR